Order Entry
Ireland
ContactUsLinkComponent
Anti-CD19 Rat Monoclonal Antibody [clone: 1D3] (FITC)
Anti-CD19 Rat Monoclonal Antibody [clone: 1D3] (FITC)
Certificates
undefined
Anti-CD19 Rat Monoclonal Antibody [clone: 1D3] (FITC)

Specifications

  • Catalog No:
  • STMC60112FI.2
  • STMC60112FI
  • STMC60112FI.1
  • Antigen name:
  • CD19
  • CD19
  • CD19
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 1D3
  • 1D3
  • 1D3
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 12478
  • 12478
  • 12478
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • IgG2a kappa

Specifications

Frequently Bought Together