Order Entry
Ireland
ContactUsLinkComponent
 
Methyl Orange TS
Catalog # CMPLJPH333
New Product
CAS Number:  
Methyl Orange TS
Catalog # CMPLJPH333
New Product
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Methyl orange TS
  • Shipping conditions:
    Store under normal laboratory conditions, at temperatures between 15...25 °C
  • Shelf life:
    24 months

Specifications

About this item

Certifications: ISO 17034, 17025, 9001 certified.