About this item
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD41, APC-conjugated.
- Megakaryocytes
- Platelets
The HIP8 antibody reacts with the ~125 kDa GPIIbα subunit of human CD41, a heterodimeric integrin expressed primarily on platelets, megakaryocytes, monocytes, and acute megakaryoblastic leukemia cells. The GPIIbα subunit is linked to a smaller ~25 kDa subunit (GPIIbβ) by a single disulfide bond. Three isoforms of CD41 that are generated by alternative splicing have been identified, one of which is expressed only by cancerous cells. CD41 associates non-covalently with CD61 (integrin b3) in a calcium-dependent manner to form the CD41/CD61 (or GPIIb/IIIa) complex, which has been identified as a receptor for several ligands, including fibrinogen, fibronectin, vitronectin, and thrombin. Binding to the CD41/CD61 complex involves an RGD tripeptidyl sequence in the ligand, and both ligand occupancy and ligand-mediated receptor clustering are functionally important in the integrin-mediated response. The HIP8 antibody reportedly blocks platelet aggregation and inhibits the activation of platelets by several compounds.
Type:
Antigen: CD41
Clonality: Monoclonal
Clone: HIP8
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Capuchin monkey, Chimpanzee, Cynomolgus monkey, Human, Pig, Rhesus monkey
Specifications
- Catalog No:
- STMC60114AZ
- STMC60114AZ.1
- Antigen name:
- CD41
- CD41
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- HIP8
- HIP8
- Reactivity:
- Baboon, Capuchin monkey, Pig, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Baboon, Capuchin monkey, Pig, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3674
- 3674
- Isotype:
- IgG1 kappa
- IgG1 kappa