Order Entry
Ireland
ContactUsLinkComponent
Anti-CD73 Mouse Monoclonal Antibody [clone: AD2] (FITC)
Anti-CD73 Mouse Monoclonal Antibody [clone: AD2] (FITC)
Certificates
undefined
Anti-CD73 Mouse Monoclonal Antibody [clone: AD2] (FITC)

Specifications

  • Catalog No:
  • STMC60044FI
  • STMC60044FI.1
  • Antigen name:
  • CD73
  • CD73
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • AD2
  • AD2
  • Reactivity:
  • Rhesus monkey, Macaque, Chimpanzee, Human
  • Rhesus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 4907
  • 4907
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together