Order Entry
Ireland
ContactUsLinkComponent
Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4] (Alexa Fluor® 488)
Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4] (Alexa Fluor® 488)
Certificates
undefined
Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60016AD.1
  • STMC60016AD
  • Antigen name:
  • CD4
  • CD4
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • OKT4
  • OKT4
  • Reactivity:
  • Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Chimpanzee, Human
  • Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 920
  • 920
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications

Frequently Bought Together