Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD54 (ICAM-1) Rat Monoclonal Antibody [clone: YN1/1.7.4]
 
undefined
Anti-CD54 (ICAM-1) Rat Monoclonal Antibody [clone: YN1/1.7.4]

 

  • Catalog No:
  • STMC60151
  • STMC60151.1
  • Antigen name:
  • CD54 (ICAM-1)
  • CD54 (ICAM-1)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • YN1/1.7.4
  • YN1/1.7.4
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 15894
  • 15894
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

 

Frequently Bought Together