Specifications
- Antigen name:CD10
- Clonality:Monoclonal
- Clone:FR4D11
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:4311
- Size:100 μg
- Pk:0,1 mL
Specifications
About this item
Mouse monoclonal IgG1 antibody against human CD10, unconjugated.
- Mammary cells
The FR4D11 antibody reacts with CD10, a 100 kDa type II transmembrane glycoprotein. CD10 is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt, and follicular germinal center lymphomas, immature and transitional B cells within adult bone marrow, and on blast cells from patients with chronic myelogenous leukemia (CML). CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts on the brush border of kidney, and gut epithelial cells. Most notably, CD10 was reported to mark human mammary bipotent progenitor cells.
Type:
Antigen: CD10
Clonality: Monoclonal
Clone: FR4D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human