Order Entry
Ireland
ContactUsLinkComponent
Benzene ≥99.5% (by GC), AnalaR NORMAPUR® ACS analytical reagent
Flammable
Health
Irritant
Benzene ≥99.5% (by GC), AnalaR NORMAPUR® ACS analytical reagent
Catalog # 21806.290
Supplier:  VWR Chemicals
New Product
CAS Number:  
Flammable
Health
Irritant
Benzene ≥99.5% (by GC), AnalaR NORMAPUR® ACS analytical reagent
Catalog # 21806.290
Supplier:  VWR Chemicals
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Purity (GC):
    min. 99.5%
  • Identity (IR):
    passes test
  • Appearance:
    clear colorless liquid
  • Coloration (APHA):
    max. 10
  • Refractive index (20°C, 589 nm):
    1.499 - 1.502
  • Alkalinity (meq/g):
    max. 0.0001
  • Acidity (meq/g):
    max. 0.0001
  • Residue on evaporation:
    max. 0.001%
  • Water :
    max. 0.05%
  • Substance darkened by H2SO4:
    passes test
  • Sulphur compounds (S):
    max. 0.005%
  • Thiophene (C4H4S):
    passes test
  • Copper (Cu):
    max. 0.00002%
  • Iron (Fe):
    max. 0.00005%
  • Lead (Pb):
    max. 0.00002%

Specification Test Results

About this item

 :
 :78.11 g/mol
Merck Index:13,01066
Boiling Pt:80,1 °C (1013 hPa)
Melting Pt:5,5 °C
Density:0,878 g/cm³ (20 °C)
Flash Pt:–11 °C
Storage Temperature:Ambient
EINECS:200-753-7
UN:1114
ADR:3,II
REACH:01-2119447106-44
 :GHS02
 :GHS08
 :GHS07
CAS Number:71-43-2
Formula:C₆H₆
MDL Number:MFCD00003009