Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD51 Mouse Monoclonal Antibody [clone: NKI-M9] (FITC)
 
undefined
Anti-CD51 Mouse Monoclonal Antibody [clone: NKI-M9] (FITC)

 

  • Catalog No:
  • STMC60043FI.1
  • STMC60043FI
  • Antigen symbol:
  • CD51
  • CD51
  • Antigen name:
  • Integrin aV
  • Integrin aV
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • NKI-M9
  • NKI-M9
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3685
  • 3685
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

 

Frequently Bought Together