Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD49f Rat Monoclonal Antibody [clone: GoH3] (APC)
 
undefined
Anti-CD49f Rat Monoclonal Antibody [clone: GoH3] (APC)

 

  • Catalog No:
  • STMC60037AZ
  • STMC60037AZ.1
  • Antigen symbol:
  • CD49f
  • CD49f
  • Antigen name:
  • Integrin a6
  • Integrin a6
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • GoH3
  • GoH3
  • Reactivity:
  • Dog, Mouse, Cat, Baboon, Capuchin monkey, Pig, Rabbit, Horse, Sheep, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Bovine
  • Dog, Mouse, Cat, Baboon, Capuchin monkey, Pig, Rabbit, Horse, Sheep, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Bovine
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16403/3655
  • 16403/3655
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

 

Frequently Bought Together