Order Entry
Ireland
ContactUsLinkComponent
Anti-PAX6 (AN) Rabbit Polyclonal Antibody [clone: Poly] ()
Anti-PAX6 (AN) Rabbit Polyclonal Antibody [clone: Poly] ()
Catalog # STMC100-1340
undefined
Anti-PAX6 (AN) Rabbit Polyclonal Antibody [clone: Poly] ()
Catalog # STMC100-1340

Specifications

  • Antigen name:
    PAX6 (AN)
  • Clonality:
    Polyclonal
  • Clone:
    Poly
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Gene ID:
    5080
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody against human, mouse PAX6 (AN).

  • Neural cells Psc derived
  • Neural stem
  • Progenitor cells

This rabbit polyclonal antibody reacts with human and mouse paired box protein Pax6, a ~46 kDa protein encoded by PAX6 gene belonging to the PAX family of transcription factors. Use this polyclonal antibody in applications requiring detection of PAX-6, such as ELISA, immunohistochemistry (IHC), western blotting (WB), immunocytochemistry (ICC), and immunofluorescence (IF). This antibody clone has been verified for labeling PAX6-positive hPSC-derived neural stem and progenitor cells generated with STEMdiff™ SMADi Neural Induction Kit (Catalog #08581) and STEMdiff™ Neural Progenitor Medium (Catalog #05833). Expressed in the brain, eyes, pancreas, and spinal cord, Pax-6 potentiates their embryonic development, as well as the maintenance of glucose homeostasis and function of the endocrine pancreas. PAX6 has at least 3 isoforms generated via alternative mRNA splicing, i.e., PAX6, PAX6(5a), and PAX6(ΔPD). PAX6 contains two DNA-binding domains, the paired domain (PD) and a paired-type homeodomain (HD) connected by a linker region. Downstream of the HD, a P/S/T-rich domain at the C-terminal acts as an activator of gene expression. The PAX6(5a) splice variant contains a 14 amino acid insert into the PD, altering the specificity of its DNA binding activity, while the PAX6(ΔPD) isoforms lack a PD, resulting in a truncated protein. In humans, mutations of the PAX6 gene resulted in aniridia and related developmental eye diseases, while Pax-6 deficiency in adult mice resulted in progressive hyperglycemia.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: PAX6 (AN)
Clonality: Polyclonal
Clone: Poly
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse