Order Entry
Ireland
ContactUsLinkComponent
 
Anti-IFN-gamma Rat Monoclonal Antibody [clone: XMG1.2] (APC)
 
undefined
Anti-IFN-gamma Rat Monoclonal Antibody [clone: XMG1.2] (APC)

 

  • Catalog No:
  • STMC100-0278
  • STMC100-0277
  • Antigen name:
  • IFN-gamma
  • IFN-gamma
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • XMG1.2
  • XMG1.2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 15978
  • 15978
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together