Order Entry
Ireland
ContactUsLinkComponent
Anti-CD123 (IL-3Rα) Mouse Monoclonal Antibody [clone: 6H6]
Anti-CD123 (IL-3Rα) Mouse Monoclonal Antibody [clone: 6H6]
Certificates
undefined
Anti-CD123 (IL-3Rα) Mouse Monoclonal Antibody [clone: 6H6]

Specifications

  • Catalog No:
  • STMC60110.1
  • STMC60110
  • Antigen name:
  • CD123 (IL-3Ra)
  • CD123 (IL-3Ra)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 6H6
  • 6H6
  • Reactivity:
  • Sooty mangabey, Rhesus monkey, Human
  • Sooty mangabey, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3563
  • 3563
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together