Order Entry
Ireland
ContactUsLinkComponent
Anti-CD20 Mouse Monoclonal Antibody [clone: 2H7]
Anti-CD20 Mouse Monoclonal Antibody [clone: 2H7]
Catalog # STMC60008
undefined
Anti-CD20 Mouse Monoclonal Antibody [clone: 2H7]
Catalog # STMC60008

Specifications

  • Antigen name:
    CD20
  • Clonality:
    Monoclonal
  • Clone:
    2H7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Baboon,
    Capuchin monkey,
    Ma's night monkey,
    Cynomolgus monkey,
    Rhesus monkey,
    Pigtailed macaque,
    Chimpanzee,
    Human,
    Squirrel monkey
  • Gene ID:
    931
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal IgG2b antibody against human, rhesus, cynomolgus CD20, unconjugated.

  • B cells

Antibody 2H7 reacts with CD20, an ~35 kDa non-glycosylated type 1 transmembrane protein in the MS4A protein family. The CD20 polypeptide transverses the cell membrane four times, with only a minor portion of the protein displayed on the cell surface. The epitope recognized by antibody 2H7 has been mapped to the amino acid sequence YNCEPANPSEKNSPST, located in the large extracellular loop of CD20. CD20 is expressed on pre-B cells, resting and activated B cells, some follicular dendritic cells, and a subset of T cells. Expression by B cells is lost upon their differentiation into plasma cells. By associating with several proteins, including the B cell receptor (CD79), MHC class I and II, CD53, CD81, and CD82, CD20 is involved in initiating intracellular signaling pathways that modulate the activation, proliferation, and differentiation of B cells. It is thought that CD20 forms multi-subunit ion channels that regulate calcium ion flux across the plasma membrane. Activation of CD20 is accompanied by pronounced phosphorylation of the cytoplasmic domain of the ~33 kDa apo-protein, with the appearance of 35 - 37 kDa isoforms that associate with Src family kinases such as Fyn, Lck and Lyn. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human Whole Blood CD20 Positive Selection Kit (Catalog #18685), EasySep™ Human CD19 Positive Selection Kit II (Catalog #17854), and EasySep™ HLA Whole Blood Lymphoid Positive Selection Kit (Catalog #18684HLA).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Baboon, Capuchin monkey, Chimpanzee, Cynomolgus monkey, Human, Ma's night monkey, Pigtailed macaque, Rhesus monkey, Squirrel monkey