Order Entry
Ireland
ContactUsLinkComponent
Anti-CD138 Mouse Monoclonal Antibody [clone: MI15]
Anti-CD138 Mouse Monoclonal Antibody [clone: MI15]
  STMC60003
undefined
Anti-CD138 Mouse Monoclonal Antibody [clone: MI15]
  STMC60003

 

  • Antigen name:
    Syndecan-1
  • Antigen symbol:
    CD138
  • Clonality:
    Monoclonal
  • Clone:
    MI15
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Cynomolgus monkey,
    Rhesus monkey,
    Human
  • Gene ID:
    6382
  • Pk:
    100 µG

 

 

Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD138 (syndecan-1), unconjugated.

  • B cells
  • Plasma

The MI15 antibody reacts with an extracellular epitope on CD138 (Syndecan-1), an ~85 - 95 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells), as well as on non-hematopoietic cells such as embryonic mesenchymal cells, endothelial, epithelial and neural cells. CD138 expression is used as a diagnostic marker for several types of tumors. CD138 is thought to act primarily as a receptor that modulates cell proliferation, cell migration and cell-matrix associations by linking the extracellular matrix to the cytoskeleton. Heparin sulfate and chondroitin sulfate moieties attached to CD138 associate with several proteins, including collagens, fibronectin, tenascin, thrombospondin and certain cytokines. The MI15 antibody recognizes a different epitope than that of the clone DL-101 anti-CD138 antibody but blocks binding of clone B-B4.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD138
Clonality: Monoclonal
Clone: MI15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cynomolgus monkey, Human, Rhesus monkey