Order Entry
Ireland
ContactUsLinkComponent
Human Recombinant IFN-gamma
Human Recombinant IFN-gamma
Certificates
undefined
Human Recombinant IFN-gamma

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    IFN-gamma
  • Purity:
    ≥95%

Specifications

Frequently Bought Together