About this item
Nerve growth factor (NGF)-beta is a prototypical member of the neurotrophin family and has a role in the survival and growth of neural cells, regulating cell growth, promoting differentiation into neurons, and neuron migration. The beta subtype of NGF is biologically active in comparison to the alpha-2 and gamma-2 subtypes. NGF-beta in its secreted form can bind to tyrosine kinase A (trkA) receptor with high affinity and to p75 (NTR) with low affinity (Levi and Alemà; Sofroniew et al.). NGF has been shown to possess pro-inflammatory and pro-fibrogenic properties (Micera et al.). It has also been shown that overexpression of NGF-beta promotes differentiation of bone marrow mesenchymal stem cells into neurons through regulation of AKT and MAPK pathways (Yuan et al.). This product is animal component-free.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Protein/peptide name:NGF-beta, ACF
- Purity:≥95%