Human Recombinant Amphiregulin
Catalog # STMC78104.1
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:1202
- Protein/peptide name:Amphiregulin
- Purity:≥95%
Specifications
About this item
Amphiregulin is a member of the epidermal growth factor (EGF) family. It binds to several receptors including EGFR/ErbB1, HER2/ErbB2, HER3/ErbB3, and HER4/ErbB4, which activates Ras/Raf/MAPK and PI3K/AKT pathways. This results in cell proliferation, apoptosis resistance, and invasive behaviour (Normanno et al.). The expression of amphiregulin is induced by tissue injury and inflammation. Amphiregulin promotes mammary development, trophoblast growth, lung and kidney branching morphogenesis, and keratinocyte proliferation. It stimulates the growth of most cell types, including normal and transformed mammary epithelial cells, and malignant cells of the colon, prostate, cervix, and pancreas (Berasain and Avila).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').