Order Entry
Ireland
ContactUsLinkComponent
JCV (LT) Peptide Pool
JCV (LT) Peptide Pool
Catalog # STMC100-1399
CAS Number:  
undefined
JCV (LT) Peptide Pool
Catalog # STMC100-1399
CAS Number:  

Specifications

  • Pk:
    1 AU
  • Seq:
    0001
  • Protein/peptide name:
    JCV (LT) Peptide Pool
  • Purity:
    Average 70%

Specifications

About this item

Stimulating T cells with JCV (LT) Peptide Pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. This peptide pool is a lyophilised mixture of 170 peptides from large T antigen (LT) of JC polyomavirus (JCV), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 688 on LT. LT is detectable during JCV latency (Rossi et al.), and in immunocompromised patients with JVC-associated pathologies such as progressive multifocal leukoencephalopathy (PML) (Delbue et al.). In PML, LT has been associated with activating the promoter of survivin, an anti-apoptotic protein (Del Valle et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').