Order Entry
Ireland
ContactUsLinkComponent
HPV16 (E6) Peptide Pool
HPV16 (E6) Peptide Pool
  STMC100-1395
 :  
undefined
HPV16 (E6) Peptide Pool
  STMC100-1395
 :  
  • Pk:
    1 AU
  • Seq:
    0001
  • Protein/peptide name:
    HPV16 (E6) Peptide Pool
  • Purity:
    Average 70%
Stimulating T cells with HPV16 (E6) Peptide Pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HPV16 (E6) Peptide Pool is a lyophilised mixture of 37 peptides from the E6 protein of human papillomavirus type 16 (HPV16). The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 158 on protein E6. E6 binds to the tumor suppressor protein p53 and stimulates its degradation (Scheffner et al.; Werness et al. ), which is thought to cause an increase in spontaneous mutagenesis in host cells (Havre et al.). E6 together with E7 demonstrated the ability to immortalize human keratinocytes in vitro, which likely has significance for HPVs oncogenic potential (Hawley-Nelson et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').