Order Entry
Ireland
ContactUsLinkComponent
Human Recombinant ANGPTL2, His Tag
Human Recombinant ANGPTL2, His Tag
Catalog # STMC100-1321
CAS Number:  
undefined
Human Recombinant ANGPTL2, His Tag
Catalog # STMC100-1321
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    1286
  • Protein synonyms:
    ARP2|HARP
  • Protein/peptide name:
    ANGPTL2
  • Purity:
    ≥92%

Specifications

About this item

Use angiopoietin-like protein 2 (ANGPTL2) to regulate tissue remodeling through integrin α5β signaling and activation of p38 mitogen-activated protein kinases (MAPK) (Odagiri et al.; Tabata et al.). Highly expressed in the heart, small intestine, and stomach, ANGPTL2 is a glycosylated secretory protein that contains a coiled domain and a fibrinogen-like domain (Kim et al.). Studies have shown that the coiled-coil domain of ANGPTL2 functions as a growth factor, enhancing the survival of hematopoietic stem cells (HSCs) ex vivo (Broxmeyer et al.). ANGPTL2 is also known to play a role in obesity and metabolic diseases, promoting local inflammation in adipose tissue and systemic insulin resistance in mice models (Tabata et al.). By activating an inflammatory cascade in endothelial cells and increasing macrophage infiltration, ANGPTL2 accelerates vascular inflammation which may lead to endothelial dysfunction and atherosclerosis progression (Horio et al.). This protein product contains a His-residue tag at the amino end of the polypeptide chain. For consistency and reproducibility across your applications, sclerostin from STEMCELL comes lyophilised with ≥92% purity, and endotoxin levels are verified to be ≤1.0 EU/μg protein.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').