Order Entry
Ireland
ContactUsLinkComponent
Human Recombinant Sclerostin, His Tag
Human Recombinant Sclerostin, His Tag
Catalog # STMC100-1299
CAS Number:  
undefined
Human Recombinant Sclerostin, His Tag
Catalog # STMC100-1299
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    1202
  • Protein synonyms:
    CDD|DAND6|SOST|SOST1|VBCH
  • Protein/peptide name:
    Sclerostin
  • Purity:
    ≥87%

Specifications

About this item

Use sclerostin to alter bone remodeling homeostasis, where it inhibits bone formation in vivo and in vitro, likely through Wnt/β-catenin signaling cascades (Ellies et al.; Lin et al.). Sclerostin is a member of the cerberus/DAN family of glycoproteins whose members share a C-terminal cysteine-knot-like (CTCK) domain. Highly expressed in bone and cartilage, as well as in kidney, and liver tissues (Weivoda et al.), this osteoclast-derived BMP antagonist binds BMP6 and BMP7 with high affinity, and binds BMP2 and BMP4 with a lower affinity (Kusu et al.). Mutations in the SOST gene have been associated with sclerosteosis (Brunkow et al.), van Buchem disease (Staehling-Hampton et al.), and bone dysplasia disorders, characterized by increased bone density. This protein product contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, sclerostin from STEMCELL comes lyophilised with ≥87% purity, and endotoxin levels are verified to be ≤1.0 EU/μg protein.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').