SARS-CoV-2 (VME1) Peptide Pool
Catalog # STMC100-0648
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:1 KIT
- Source:
- Species:
- Seq:0075
- Environmentally Preferable:
- Protein/peptide name:SARS-CoV-2 (VME1) Peptide Pool
- Purity:Average 70%
Specifications
About this item
The SARS-CoV-2 (VME1) peptide pool is a lyophilised mixture of 53 peptides from the SARS-CoV-2 membrane protein VME1. VME1 is a structural protein that is common to all coronaviruses, and it plays a critical role in viral assembly. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 222 on VME1.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').