Order Entry
Ireland
Orders LinkContactUsLinkComponent
Human Recombinant IL-15

Human Recombinant IL-15

Marketsource
Human Recombinant IL-15
Marketsource
Certificates
Human Recombinant IL-15

3 Options Available Below

Product Details & Documents

Interleukin 15 (IL-15) is a four-alpha helix bundle cytokine with many similar properties to IL-2, with which it shares components of its receptor. The IL-15 receptor is a heterotrimeric receptor composed of IL-15Ra, the high-affinity receptor for IL-15, as well as IL-2/15Rb (CD122) and common gamma chain (CD132). IL-15 binds to IL-15Rα receptor and can then be presented in trans to IL-2/15Rb and common gamma chain on other cells. Trans-presentation is thought to be the major mechanism by which IL-15-mediated responses occur in mice, although may not be necessary in humans (Castillo et al.). The cytoplasmic domains of IL-2/15Rb and common gamma chain mediate signaling to activate JAK/STAT and PI3K pathways. IL-15 supports the survival and proliferation of naive CD4+ and CD8+ T cells, and promotes homeostasis of memory T cells. IL-15 also promotes the survival and differentiation of NK cells and regulates their cytolytic activity (Ma et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • STMC78031.3
  • STMC78031.1
  • STMC78031
  • Protein/peptide name:
  • IL-15
  • IL-15
  • IL-15
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1.000 µG
    Log in for availability
    Log in for availability
    1 * 1.000 µG
    1 * 1.000 µG - 5 440,00 €
    5 440,00 €
    1 * 1.000 µG
       5 440,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78031.3
    Protein/peptide name
    IL-15
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
  • 10 µG
    Log in for availability
    Log in for availability
    1 * 10 µG
    1 * 10 µG - 275,00 €
    275,00 €
    1 * 10 µG
       275,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78031.1
    Protein/peptide name
    IL-15
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
  • 100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 1 090,00 €
    1 090,00 €
    1 * 100 µG
       1 090,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78031
    Protein/peptide name
    IL-15
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource