Order Entry
Ireland
ContactUsLinkComponent
HHV6 (U90) Peptide Pool
HHV6 (U90) Peptide Pool
Catalog # STMC100-1402
CAS Number:  
undefined
HHV6 (U90) Peptide Pool
Catalog # STMC100-1402
CAS Number:  

Specifications

  • Pk:
    1 AU
  • Protein/peptide name:
    HHV6 (U90) Peptide Pool
  • Purity:
    Average 70%

Specifications

About this item

Stimulating T cells with HHV6 (U90) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HHV6 (U90) Peptide pool is a lyophilised mixture of 267 peptides from protein U90 of human herpesvirus 6B (HHV-6B; strain Z29) and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 1078 on U90. U89 - U90 RNA expression occurs under immediate-early conditions of infection (Mirandola et al.), and the associated protein exhibits peri-nuclear and cytoplasmic localization (Nikolaou et al.). Also known as immediate-early protein IE1, latency-associated transcripts of HHV6 may function as a source of IE1 during viral reactivation (Kondo et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').