FIM-1, Sigma-Aldrich®
Catalog # F9926-250UG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:250 µG
- Seq:1370
Specifications
About this item
:
:755.75 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
CAS Number:220518-50-3
Formula:C₄₅H₃₁N₄O₈
MDL Number:MFCD03453075