Order Entry
Ireland
ContactUsLinkComponent
 
Irritant
4-Amino-4'-dimethylaminoazobenzene 97%, Sigma-Aldrich®
Catalog # 379298-1G
CAS Number:  
undefined
Irritant
4-Amino-4'-dimethylaminoazobenzene 97%, Sigma-Aldrich®
Catalog # 379298-1G
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance (Colour):
    Orange
  • Appearance (Form), Powder, Crystals and/or Chunks:
    Conforms to Requirements
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    67.5 - 72.4%
  • Nitrogen:
    22.5 - 24.1%
  • Purity (HPLC):
    ≥96.5%

Specification Test Results

About this item

 :
 :240.31 g/mol
Melting Pt:190 °C (dec., lit.)
EINECS:208-712-5
 :GHS07
Storage Temperature:Room Temperature
CAS Number:539-17-3
Formula:C₁₄H₁₆N₄
MDL Number:MFCD00010204