Hexaamminecobalt trichloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride
CAS Number:10534-89-1
:
Formula:H₁₈CoCl₃N₆
MDL Number:MFCD00036304
:267.47 g/mol
Merck Index:14,4675
Density:1,71
EINECS:234-103-9
:GHS08
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together