Hexaamminecobalt trichloride 99%
: Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride
:10534-89-1
:
:H₁₈CoCl₃N₆
:MFCD00036304
:267.47 g/mol
:217 °C
:12,04711
:234-103-9
:GHS08
:Ambient
- Environmentally Preferable:
- Appearance:Orange to orange-brown Powder
- Titration Argentometric:≥98.5 %
- Refractive index:1.4170 to 1.4240 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together