Order Entry
Ireland
ContactUsLinkComponent
Aroclor 1221 (PCB-mixture, 21% chlorinated)
Aroclor 1221 (PCB-mixture, 21% chlorinated)
Catalog # ACCUC-221N
Supplier:  AccuStandard
CAS Number:  
Aroclor 1221 (PCB-mixture, 21% chlorinated)
Catalog # ACCUC-221N
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
Boiling Pt:275 °C (1013 hPa)
Density:1,4 g/cm³ (20 °C)
UN:2315
ADR:9,II
CAS Number:11104-28-2
 :1336-36-3