Aroclor 1016 (PCB-mixture, 42% chlorinated)
Catalog # ACCUC-216N
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
Boiling Pt:385 °C (1013 hPa)
Density:1,4 g/cm³ (20 °C)
UN:2315
ADR:9,II
CAS Number:12674-11-2
:1336-36-3