
Eosin Y (yellowish), Certistain® for microscopy, Sigma-Aldrich®
Catalog # 1.15935.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:300 g/l (20 °C)
- Melting Point:295 - 296 °C
- Molar Mass:691.86 g/mol
- Bulk Density:710 kg/m3
- pH:9.2 (10 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
Merck Index:13,03634
Melting Pt:295…296 °C
:M-Clarity™ quality level = MQ300
:691.86 g/mol
EINECS:241-409-6
:GHS07
Storage Temperature:Ambient
:1
CAS Number:17372-87-1
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040