
Fuchsin (acid), pure, high purity biological stain
:3244-88-0
:
:C₂₀H₁₇N₃Na₂O₉S₃
:MFCD00013286
:585.55 g/mol
:12,00109
:? 130 °C
:221-816-5
:GHS07
:Ambient
- Appearance:Dark green Powder or crystals
- Infrared spectrum:Conforms
- UV Lambda max:544 to 550 nm (in pH 3 acidified water)
- Absorption Ratio P-15/P+15:1 to 1.4
- Specific absorbance A (1%/1cm):≥700 (at lambda max.)
Frequently Bought Together