Thiamine hydrochloride, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:67-03-8
:
Formula:C₁₂H₁₈Cl₂N₄OS
MDL Number:MFCD00012780
:337.27 g/mol
Merck Index:13,09366
Melting Pt:252 °C
Density:1,401 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-641-8
Frequently Bought Together