Order Entry
Ireland
ContactUsLinkComponent
 
Stainless Steel Tray
  SAMPA236-500
undefined
Stainless Steel Tray
  SAMPA236-500

 

  • Description:
    Deep tray
  • Material:
    Stainless steel 316L
  • W×D×H:
    535×330×95 mm
  • Pk:
    1

 

 

Designed for heavy-duty use in pharmaceutical, chemical, and food facilities.

  • Made from high-quality 316L stainless steel
  • Mirror polish finish and radiused corners for easy cleaning
  • Available from stock
  • Fast delivery

316L stainless steel trays are perfect for heavy-duty use across a range of industries, including food and pharmaceutical. Design features in line with GMP guidelines ensure they are easy to both use and clean.