IP-10 (from E. coli)
US-made bioactive recombinant proteins offer rigid quality testing and low manufacturer pricing.
- Strict quality standards
Interferon gamma-induced protein 10 (IP-10), or CXCL10, is a chemokine secreted by monocytes, endothelial cells and fibroblasts in response to interferon gamma (IFN-ɣ). IP-10 functions as a chemoattractant for activated T cells, monocytes, dendritic, and natural killer (NK) cells that express the G protein-coupled receptor CXCR3. IP-10 is an important factor in autoimmune diseases such as Hashimoto's thyroiditis, Graves' disease, and Type 1 diabetes mellitus.
US-made bioactive recombinant proteins offer rigid quality testing and low manufacturer pricing.
- Strict quality standards
Interferon gamma-induced protein 10 (IP-10), or CXCL10, is a chemokine secreted by monocytes, endothelial cells and fibroblasts in response to interferon gamma (IFN-ɣ). IP-10 functions as a chemoattractant for activated T cells, monocytes, dendritic, and natural killer (NK) cells that express the G protein-coupled receptor CXCR3. IP-10 is an important factor in autoimmune diseases such as Hashimoto's thyroiditis, Graves' disease, and Type 1 diabetes mellitus.
- Catalog No:
- SHBT400-02-5UG
- SHBT400-02-25UG
- SHBT400-02-100UG
- SHBT400-02-500UG
- SHBT400-02-1MG
- Protein/peptide name:
- IP-10
- IP-10
- IP-10
- IP-10
- IP-10
- Protein synonyms:
- CXCL10|CRG-2
- CXCL10|CRG-2
- CXCL10|CRG-2
- CXCL10|CRG-2
- CXCL10|CRG-2
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Guinea pig
- Guinea pig
- Guinea pig
- Guinea pig
- Guinea pig
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- H0UXM5
- H0UXM5
- H0UXM5
- H0UXM5
- H0UXM5
- Sequence:
- IPHSRTIRCT CIETSTQPVN PKSFKKLEII PASQSCPRVE IIATMKMNGE KRCLDPESKV IKNLLKAVRK ERSKRS
- IPHSRTIRCT CIETSTQPVN PKSFKKLEII PASQSCPRVE IIATMKMNGE KRCLDPESKV IKNLLKAVRK ERSKRS
- IPHSRTIRCT CIETSTQPVN PKSFKKLEII PASQSCPRVE IIATMKMNGE KRCLDPESKV IKNLLKAVRK ERSKRS
- IPHSRTIRCT CIETSTQPVN PKSFKKLEII PASQSCPRVE IIATMKMNGE KRCLDPESKV IKNLLKAVRK ERSKRS
- IPHSRTIRCT CIETSTQPVN PKSFKKLEII PASQSCPRVE IIATMKMNGE KRCLDPESKV IKNLLKAVRK ERSKRS
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Form:
- Lyophilised
- Lyophilised
- Lyophilised
- Lyophilised
- Lyophilised
- Formulation:
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- Molecular weight:
- 8,7 kDa
- 8,7 kDa
- 8,7 kDa
- 8,7 kDa
- 8,7 kDa
- Reconstitution instructions:
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
Product Family Options
- Pack typePk
Specifications
Cat. No.SHBT400-02-5UGProtein/peptide nameIP-10Protein synonymsCXCL10|CRG-2Protein/peptide typeRecombinantProtein functionCytokineSpeciesGuinea pigSourceE. coliConjugationUnconjugatedGene IDH0UXM5SequenceIPHSRTIRCT CIETSTQPVN PKSFKKLEII PASQSCPRVE IIATMKMNGE KRCLDPESKV IKNLLKAVRK ERSKRSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormLyophilisedFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight8,7 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTSpecifications
Cat. No.SHBT400-02-25UGProtein/peptide nameIP-10Protein synonymsCXCL10|CRG-2Protein/peptide typeRecombinantProtein functionCytokineSpeciesGuinea pigSourceE. coliConjugationUnconjugatedGene IDH0UXM5SequenceIPHSRTIRCT CIETSTQPVN PKSFKKLEII PASQSCPRVE IIATMKMNGE KRCLDPESKV IKNLLKAVRK ERSKRSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormLyophilisedFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight8,7 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTSpecifications
Cat. No.SHBT400-02-100UGProtein/peptide nameIP-10Protein synonymsCXCL10|CRG-2Protein/peptide typeRecombinantProtein functionCytokineSpeciesGuinea pigSourceE. coliConjugationUnconjugatedGene IDH0UXM5SequenceIPHSRTIRCT CIETSTQPVN PKSFKKLEII PASQSCPRVE IIATMKMNGE KRCLDPESKV IKNLLKAVRK ERSKRSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormLyophilisedFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight8,7 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTSpecifications
Cat. No.SHBT400-02-500UGProtein/peptide nameIP-10Protein synonymsCXCL10|CRG-2Protein/peptide typeRecombinantProtein functionCytokineSpeciesGuinea pigSourceE. coliConjugationUnconjugatedGene IDH0UXM5SequenceIPHSRTIRCT CIETSTQPVN PKSFKKLEII PASQSCPRVE IIATMKMNGE KRCLDPESKV IKNLLKAVRK ERSKRSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormLyophilisedFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight8,7 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTSpecifications
Cat. No.SHBT400-02-1MGProtein/peptide nameIP-10Protein synonymsCXCL10|CRG-2Protein/peptide typeRecombinantProtein functionCytokineSpeciesGuinea pigSourceE. coliConjugationUnconjugatedGene IDH0UXM5SequenceIPHSRTIRCT CIETSTQPVN PKSFKKLEII PASQSCPRVE IIATMKMNGE KRCLDPESKV IKNLLKAVRK ERSKRSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormLyophilisedFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight8,7 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



