Naphthol AS-GR phosphate disodium salt
Catalog # N3625-1G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Clear to slightly hazy dark yellow solution at 50 mg/ml of water
- Solubility:Conforms to structure
- IR Spectrum:< or = 23% (7 Mol/Mol)
- Water by Karl Fischer:6.6 to 7.1% Phosphorus
- ICP-Atomic Emission:Jun-03
- Purity by thin layer chromatography:* Anhydrous Basis
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ200
:451.32 g/mol
Storage Temperature:Freezer
CAS Number:100929-51-9
Formula:C₂₂H₁₆NO₅PNa₂
MDL Number:MFCD00069449