Order Entry
Ireland
ContactUsLinkComponent
10% Bovine Serum Albumin in Iscove's MDM
10% Bovine Serum Albumin in Iscove's MDM
Catalog # STMC09300
CAS Number:  
undefined
10% Bovine Serum Albumin in Iscove's MDM
Catalog # STMC09300
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    10% bovine serum albumin in Iscove's MDM

Specifications

About this item

10% BSA in Iscove's MDM.

This product has been developed for use in applications where a culture medium of defined composition is required. It contains pre-screened batches of bovine serum albumin (BSA) that have been selected to support the optimal growth of human hematopoietic progenitor cells in serum-free media formulations. It is also suitable for the culture of mouse hematopoietic progenitor cells in serum-free conditions. Recommended for use with MethoCult™ H4100 or MethoCult™ M3134 for hematopoietic colony-forming unit (CFU) assays (at a final concentration of 1%).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').