Order Entry
Ireland
ContactUsLinkComponent
Iscove's MDM with 2% FBS
Iscove's MDM with 2% FBS
  STMC07700
 :  
undefined
Iscove's MDM with 2% FBS
  STMC07700
 :  

 

  • Pk:
    100 mL
  • Description:
    Iscove's MDM with 2% FBS

 

 

Medium for preparing and washing samples for CFU assays.

Iscove's Modified Dulbecco's Medium (MDM) with 2% Fetal Bovine Serum (FBS) is recommended for preparing and washing human cord blood, peripheral blood, or bone marrow samples for colony-forming unit (CFU) assays.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').