MesenCult™ MSC Basal Medium (Human)
STMC05401
:
- Pk:450 mL
- Description:MesenCult™ MSC basal medium (Human)
Basal medium for human mesenchymal stem cells.
MesenCult™ MSC basal medium (Human) is a standardised basal medium designed to be supplemented with MesenCult™ mesenchymal stem cell stimulatory supplement (Human) for the in vitro culture of human mesenchymal stem cells (MSCs). MesenCult™ MSC basal medium is a component of MesenCult™ proliferation kit (Human), and is also available separately.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').