Order Entry
Ireland
ContactUsLinkComponent
TeSR™ E8™ 3D Culture Medium
TeSR™ E8™ 3D Culture Medium
Catalog # STMC03990
CAS Number:  
undefined
TeSR™ E8™ 3D Culture Medium
Catalog # STMC03990
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    TeSR™ E8™ 3D
Specifications
About this item
Animal component-free suspension culture medium for human ES and iPS cells.
TeSR™-E8™ 3D is an animal component-free culture medium developed for the expansion and scale-up of undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells as aggregates in 3D suspension culture. Based on TeSR™-E8™, TeSR™-E8™3D is a low-protein medium with an optimised protocol for scaling up human ES and iPS cell production in suspension culture. The novel fed-batch workflow saves time and media; daily feeds replenish nutrients and eliminate the need for medium exchanges on non-passaging days. TeSR™-E8™3D is made with the highest level of quality and care. It contains only the essential components required for scale-up of ES and iPS cells. The TeSR™-E8™3D system is compatible with cells previously cultured in mTeSR™1 or TeSR™-E8™ on Corning® Matrigel® hESC-qualified matrix or Vitronectin XF™, a matrix developed and manufactured by nucleus biologics), and is compatible with a range of suspension culture vessels.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').