Sodium borate solution 5 g/dl in deionised water
Catalog # HT1002-100ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance (Colour):Colorless
- Appearance (Form):Solution
- Appearance (Turbidity)
Essentially free of particles/precipitate:Clear - Performance
When using Sigma-Aldrich Procedure No. HT100, fungi organisms stain purplish-brown to black:Conforms - pH:8.9 - 9.6
- Expiration Date Period:24 months
Specification Test Results
About this item
:
:GHS08
Storage Temperature:Room Temperature
:M-Clarity™ quality level = MQ500 (more info.)
MDL Number:MFCD00081185