Haematoxylin, Sigma-Aldrich®
Catalog # GHS1128-4L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Appearance (Turbidity):Clear
- Appearance(Colour)
Dark Reddish-Purple:Conforms - Solution:Conforms
- undefined:Appearance (Form)
- Conforms:Essentially free of precipitate
- 24 months:Using Sigma-Aldrich Procedure No. GHS, Nuclei and compontents stain blue to black. Chromatin is clearly visible.
Specification Test Results
About this item
:
:302.28 g/mol
:M-Clarity™ quality level = MQ500 (more info.)
:GHS07
:GHS08
Storage Temperature:Room Temperature
CAS Number:517-28-2
:208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111