6-Aminofluorescein (isomer II), Sigma-Aldrich®
Catalog # 201634-1G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance (Colour):Yellow to Orange
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Carbon:65.4 - 72.9%
- Nitrogen:3.7 - 4.3%
- Water (by Karl Fischer):≤5.0%
- Purity (HPLC):≥90.0%
- Solubility (Turbidity), c = 1 mg/ml (0.1%), Methanol:Clear
- Solubility (Colour), Dark Yellow to Orange:Conforms to Requirements
Specification Test Results
About this item
:
:347.33 g/mol
Melting Pt:285 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
EINECS:257-334-7
CAS Number:51649-83-3
Formula:C₂₀H₁₃NO₅
MDL Number:MFCD00005051