



Mesitylene ≥98.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:108-67-8
:
Formula:C₉H₁₂
MDL Number:MFCD00008538
Merck Index:13,05934
Boiling Pt:165 °C (1013 hPa)
Melting Pt:–45 °C
Density:0,8637 g/cm³ (20 °C)
Flash Pt:48 °C
UN:2325
ADR:3,III
:120.19 g/mol
EINECS:203-604-4
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay:≥ 98,0 %
- Identification (IR):Passes test
- Density (20/4):0,864 - 0,866
Specification Test Results
Related Information
Frequently Bought Together