Anti-CD8, alpha chain Mouse Monoclonal Antibody [clone: SK1] (mFluor™ Violet 500)
Supplier: AAT Bioquest
Certificates
About this item
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.
- Affinity purified
- Easily reconstituted
Caution: For research use only.
Type: Primary
Antigen: CD8, alpha chain
Clonality: Monoclonal
Clone: SK1
Conjugation: mFluor™ Violet 500
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB10081101
- AATB10081100
- Antigen symbol:
- CD8, alpha chain
- CD8, alpha chain
- Antigen name:
- CD8 antigen, alpha chain
- CD8 antigen, alpha chain
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- mFluor™ Violet 500
- mFluor™ Violet 500
- Clone:
- SK1
- SK1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- Freeze (<–15 °C); Minimise light exposure
- Freeze (<–15 °C); Minimise light exposure
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together