Order Entry
Ireland
ContactUsLinkComponent
Anti-PURG Rabbit Polyclonal Antibody
Anti-PURG Rabbit Polyclonal Antibody
Catalog # PRSI55-290
Supplier:  ProSci Inc.
New Product
undefined
Anti-PURG Rabbit Polyclonal Antibody
Catalog # PRSI55-290
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Purine-rich element-binding protein gamma
  • Antigen symbol:
    PURG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Amino Acid Range = 225-253
  • Form:
    Liquid
  • Gene ID:
    Q9UJV8
  • Antigen synonyms:
    PURG
  • Modification:
    None
  • Storage buffer:
    Supplied in PBS with 0,09% (W/V) sodium azide
  • Molecular weight:
    40 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This PURG antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 225-253 amino acids from the C-terminal region of human PURG.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification.
  • Size:
    400 µl
  • Pk:
    400 µl

Specifications

About this item

The exact function of this gene is not known, however, its encoded product is highly similar to purine-rich element binding protein A. The latter is a DNA-binding protein which binds preferentially to the single strand of the purine-rich element termed PUR, and has been implicated in the control of both DNA replication and transcription. PURG lies in close proximity to the Werner syndrome gene, but on the opposite strand, on chromosome 8p11. Two transcript variants encoding different isoforms have been found for this gene.

For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: PURG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human