Order Entry
Ireland
ContactUsLinkComponent
Anti-MB21D1 Rabbit Polyclonal Antibody
Anti-MB21D1 Rabbit Polyclonal Antibody
Catalog # PRSI55-229
Supplier:  ProSci Inc.
undefined
Anti-MB21D1 Rabbit Polyclonal Antibody
Catalog # PRSI55-229
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclic GMP-AMP synthase
  • Antigen symbol:
    MB21D1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    Amino Acid Range = 266-295
  • Form:
    Liquid
  • Gene ID:
    Q8N884
  • Antigen synonyms:
    MB21D1
  • Modification:
    None
  • Storage buffer:
    Supplied in PBS with 0,09% (W/V) sodium azide
  • Molecular weight:
    59 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    0,3 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This CF150 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 266-295 amino acids from the Central region of human CF150.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification.
  • Size:
    400 µl
  • Pk:
    400 µl

Specifications

About this item

The exact function of C6orf150 remains unknown.

For FACS starting dilution is: 1:25 For WB starting dilution is: 1:500-1:1000.

Type: Primary
Antigen: MB21D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human