Specifications
- Antibody type:Primary
- Antigen name:Carbonic anhydrase 9
- Antigen symbol:CA9
- Clonality:Monoclonal
- Clone:SPM314
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q16790
- Antigen synonyms:CA9
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:The microsomal fraction of human renal cortical tissue homogenate was used as the immunogen for this CA9 antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recognises a glycoprotein of ~200 kDa, identified as carbonic anhydrase IX (CAIX/CA9/gp200). Its epitope resides in the carbohydrate domain of gp200. It shows no significant cross-reactivity with other carbohydrate determinants, such as the Lewis blood group antigens, epithelial membrane antigen, HMFG, and AB blood group antigens. In normal kidney, CA9 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Reportedly, CA9 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas. This mAb may be useful in the investigations of carcinomas of proximal nephrogenic differentiation especially those showing tubular differentiation.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the CA9 antibody for each application should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Citrate Buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: SPM314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human, Horse