Specifications
- Antibody type:Primary
- Antigen name:asparaginase like 1
- Antigen symbol:ASRGL1
- Clonality:Monoclonal
- Clone:CRASH/1289
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q7L266
- Antigen synonyms:ASRGL1
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:A human partial recombinant protein was used as the immunogen for this ASRGL1 antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ASRGL1 (Asparaginase-like protein 1), also known as CRASH, belongs to the Ntn-hydrolase family. Asparaginases utilize asparagine as a substrate to produce aspartic acid and ammonia. ASRGL1 has been identified as a autoantigenic protein that is present in the mid-piece of sperm after obstruction of the male reproductive tract. It is expressed highly in testis, but is also expressed in brain, kidney and gastrointestinal tissues. High levels of ASRGL1 have also been identified in ovarian, uterine and mammary tumors in comparison with normal tissues of the same origin.
Western blot: 0,5 - 1 µg/ml Flow Cytometry: 0,5 - 1 µg/10^6 cells in 0,1 ml IF: 0,5 - 1 µg/ml IHC (FFPE): 0,5 - 1 µg/ml The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the ASRGL1 antibody to be titered up or down for optimal performance.
Type: Primary
Antigen: ASRGL1
Clonality: Monoclonal
Clone: CRASH/1289
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human